Learn More
Molecular Devices Molecular Devices FLIPR Calcium Assay Kit, Express, Fluorescent, 96/384/1536-Plate
Catalog No. 5000014559
Encompass_Preferred
The FLIPR Calcium 6 Express Kit is a fluorescence-based, homogeneous reagent set for detecting intracellular calcium flux in high-throughput screening. The kit delivers an enhanced signal window and improved resistance to organic anion transporters, reducing the need for probenecid and enabling robust GPCR and ion channel assays across 96-, 384-, and 1536-well formats.
  • Enhanced signal window for improved assay sensitivity.
  • Homogeneous fluorescence format for simplified workflows.
  • Reduced need for probenecid due to dye resistance to organic anion transporters.
  • Supplies sufficient reagent for approximately 100 96- or 384-well plates, or 150 1536-well plates.
  • Store reagents at -20°C; aliquots may be frozen for short-term use.
Catalog No. 50-000-14559 Supplier Molecular Devices Supplier No. R8195
Add to Cart
Add to Cart
Request a Quote
The FLIPR Calcium 6 Express Kit is a fluorescence-based, homogeneous reagent set for detecting intracellular calcium flux in high-throughput screening. The kit delivers an enhanced signal window and improved resistance to organic anion transporters, reducing the need for probenecid and enabling robust GPCR and ion channel assays across 96-, 384-, and 1536-well formats.
  • Enhanced signal window for improved assay sensitivity.
  • Homogeneous fluorescence format for simplified workflows.
  • Reduced need for probenecid due to dye resistance to organic anion transporters.
  • Supplies sufficient reagent for approximately 100 96- or 384-well plates, or 150 1536-well plates.
  • Store reagents at -20°C; aliquots may be frozen for short-term use.