Learn More
Molecular Devices Molecular Devices Calcium 6 Assay Kit, Bulk, 100 Plates, 96/384/1536-Well
Catalog No. 5000014347
Encompass_Preferred
The FLIPR Calcium 6 Bulk Kit is a reagent set formulated for high-throughput calcium flux assays, supplied in a bulk format to support large screening campaigns. The kit provides multiple vials of the primary dye component and is compatible with common microplate formats and FLIPR/FlexStation readers.
  • Bulk format for 100 plates.
  • Includes 10 vials of Component A; each vial sufficient for multiple plates.
  • Compatible with 96-well, 384-well, and 1536-well plate formats.
  • Assay buffer (Component B) not included; requires separate preparation or purchase.
  • Optimized for calcium flux assays on plate readers and high-throughput screening systems.
Catalog No. 50-000-14347 Supplier Molecular Devices Supplier No. R8191
Add to Cart
Add to Cart
Request a Quote
The FLIPR Calcium 6 Bulk Kit is a reagent set formulated for high-throughput calcium flux assays, supplied in a bulk format to support large screening campaigns. The kit provides multiple vials of the primary dye component and is compatible with common microplate formats and FLIPR/FlexStation readers.
  • Bulk format for 100 plates.
  • Includes 10 vials of Component A; each vial sufficient for multiple plates.
  • Compatible with 96-well, 384-well, and 1536-well plate formats.
  • Assay buffer (Component B) not included; requires separate preparation or purchase.
  • Optimized for calcium flux assays on plate readers and high-throughput screening systems.