Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Devices Molecular Devices Calcium Assay Kit, QF, Express, 96/384/1536-well

Catalog No. 5000014652
Encompass_Preferred

The FLIPR Calcium 6-QF Express Kit from Molecular Devices is a fluorescence-based, homogeneous calcium indicator assay designed for high-throughput intracellular calcium flux screening. The QF formulation supports quench-sensitive assays and multiplexed workflows, and the Express Kit supplies reagent vials sufficient for multiple microplates in 96-, 384-, and 1536-well formats.

  • Homogeneous fluorescence-based calcium flux assay for cell-based screening.
  • QF formulation for quench-sensitive targets and multiplexing.
  • Contains 2 vials of Component A and 2 vials of Component C.
  • Assay buffer (Component B) is not included in the Express Kit.
  • Sufficient reagent for up to 100 96- or 384-well plates, or 150 1536-well plates.
  • Suited for GPCR, ion channel, and other calcium signaling assays in high-throughput screening.

Catalog No. 50-000-14652 Supplier Molecular Devices Supplier No. R8196
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The FLIPR Calcium 6-QF Express Kit from Molecular Devices is a fluorescence-based, homogeneous calcium indicator assay designed for high-throughput intracellular calcium flux screening. The QF formulation supports quench-sensitive assays and multiplexed workflows, and the Express Kit supplies reagent vials sufficient for multiple microplates in 96-, 384-, and 1536-well formats.

  • Homogeneous fluorescence-based calcium flux assay for cell-based screening.
  • QF formulation for quench-sensitive targets and multiplexing.
  • Contains 2 vials of Component A and 2 vials of Component C.
  • Assay buffer (Component B) is not included in the Express Kit.
  • Sufficient reagent for up to 100 96- or 384-well plates, or 150 1536-well plates.
  • Suited for GPCR, ion channel, and other calcium signaling assays in high-throughput screening.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.