Learn More
Molecular Devices Molecular Devices Assay Kit, Calcium 6, Evaluation, Fluorescence, 96/384/1536-well
Catalog No. 5000014343
Encompass_Preferred
The FLIPR Calcium 6 Evaluation Kit provides a homogeneous, fluorescence-based method to detect intracellular calcium changes in cell-based assays. It is formulated for use with compatible fluorescence plate readers and supports multiple microplate formats.
  • Homogeneous fluorescence assay for intracellular calcium detection.
  • Evaluation kit sized for three assay plates and three QF assay plates.
  • Includes 3 vials of component A, 3 vials of component C for QF, and 1 bottle of assay buffer.
  • Compatible with 96-well, 384-well, and 1536-well microplates.
  • Designed for screening GPCRs, ion channels, and calcium-mediated signaling.
  • Each reagent vial is sufficient for one plate, depending on dispenser dead volume.
Catalog No. 50-000-14343 Supplier Molecular Devices Supplier No. R8194
The FLIPR Calcium 6 Evaluation Kit provides a homogeneous, fluorescence-based method to detect intracellular calcium changes in cell-based assays. It is formulated for use with compatible fluorescence plate readers and supports multiple microplate formats.
  • Homogeneous fluorescence assay for intracellular calcium detection.
  • Evaluation kit sized for three assay plates and three QF assay plates.
  • Includes 3 vials of component A, 3 vials of component C for QF, and 1 bottle of assay buffer.
  • Compatible with 96-well, 384-well, and 1536-well microplates.
  • Designed for screening GPCRs, ion channels, and calcium-mediated signaling.
  • Each reagent vial is sufficient for one plate, depending on dispenser dead volume.