Learn More
Molecular Devices Molecular Devices Fura-2 QBT Calcium Kit, Explorer, 10-Plate, 96/384/1536-Well
Catalog No. 5000014660
Encompass_Preferred
The Fura-2 QBT Calcium Explorer Kit from Molecular Devices is a homogeneous, ratiometric assay kit for plate-based calcium measurements. The kit uses a quench-based format with a Fura-2 indicator to enable no-wash assays and reduce cell-based variability.
  • Homogeneous ratiometric calcium assay that eliminates wash steps.
  • Explorer format contains reagents sufficient for ten microplates.
  • Each vial of Component A is sufficient for one microplate.
  • Compatible with 96-well, 384-well, and 1536-well plate formats.
  • Uses a ratiometric Fura-2 indicator with ultraviolet excitation to reduce fluorescent interference.
Catalog No. 50-000-14660 Supplier Molecular Devices Supplier No. R8197
The Fura-2 QBT Calcium Explorer Kit from Molecular Devices is a homogeneous, ratiometric assay kit for plate-based calcium measurements. The kit uses a quench-based format with a Fura-2 indicator to enable no-wash assays and reduce cell-based variability.
  • Homogeneous ratiometric calcium assay that eliminates wash steps.
  • Explorer format contains reagents sufficient for ten microplates.
  • Each vial of Component A is sufficient for one microplate.
  • Compatible with 96-well, 384-well, and 1536-well plate formats.
  • Uses a ratiometric Fura-2 indicator with ultraviolet excitation to reduce fluorescent interference.