Learn More
Molecular Devices Molecular Devices Assay Kit, Calcium 6-QF, Quench-Free, Bulk, 100 Plates
Catalog No. 5000014659
Encompass_Preferred
The FLIPR Calcium 6-QF bulk assay kit is a quench-free, fluorescence-based reagent set for measuring intracellular calcium flux in high-throughput screening. The QF formulation is optimized for quench-sensitive targets and enables multiplexing with luminescent assays while minimizing probenecid use.
  • Quench-free formulation suitable for quench-sensitive assays and multiplexing.
  • Bulk kit sufficient for 100 96-well or 384-well plates, or 150 1536-well plates.
  • Includes 10 vials of Component A and 10 vials of Component C, each vial covering multiple plates.
  • Assay buffer not included; HBSS plus 20 mM HEPES is recommended as a substitute.
  • Compatible with high-throughput FLIPR and FlexStation microplate readers.
  • Store reagents at -20°C and other components at 4°C; reagents stable for six months in original packaging.
Catalog No. 50-000-14659 Supplier Molecular Devices Supplier No. R8193
Add to Cart
Add to Cart
Request a Quote
The FLIPR Calcium 6-QF bulk assay kit is a quench-free, fluorescence-based reagent set for measuring intracellular calcium flux in high-throughput screening. The QF formulation is optimized for quench-sensitive targets and enables multiplexing with luminescent assays while minimizing probenecid use.
  • Quench-free formulation suitable for quench-sensitive assays and multiplexing.
  • Bulk kit sufficient for 100 96-well or 384-well plates, or 150 1536-well plates.
  • Includes 10 vials of Component A and 10 vials of Component C, each vial covering multiple plates.
  • Assay buffer not included; HBSS plus 20 mM HEPES is recommended as a substitute.
  • Compatible with high-throughput FLIPR and FlexStation microplate readers.
  • Store reagents at -20°C and other components at 4°C; reagents stable for six months in original packaging.