Learn More
Molecular Devices Molecular Devices Neurotransmitter Uptake Assay, Fluorescent, Homogeneous, 96/384-Well
Catalog No. 5000014353
Encompass_Preferred
The Molecular Devices Neurotransmitter Transporter Uptake Assay Kit is a homogeneous, mix-and-read fluorescent assay for measuring live-cell uptake activity of serotonin, norepinephrine, and dopamine transporters. It supports kinetic and endpoint workflows and is suitable for high-throughput screening and assay development.
  • Homogeneous mix-and-read fluorescent format compatible with automation.
  • Suitable for kinetic real-time measurements and endpoint assays.
  • Compatible with 96-well and 384-well plate formats.
  • Evaluation kit configuration includes reagents sufficient for two plates.
  • Replaces radioactive uptake assays, reducing disposal and safety concerns.
Catalog No. 50-000-14353 Supplier Molecular Devices Supplier No. R6138
The Molecular Devices Neurotransmitter Transporter Uptake Assay Kit is a homogeneous, mix-and-read fluorescent assay for measuring live-cell uptake activity of serotonin, norepinephrine, and dopamine transporters. It supports kinetic and endpoint workflows and is suitable for high-throughput screening and assay development.
  • Homogeneous mix-and-read fluorescent format compatible with automation.
  • Suitable for kinetic real-time measurements and endpoint assays.
  • Compatible with 96-well and 384-well plate formats.
  • Evaluation kit configuration includes reagents sufficient for two plates.
  • Replaces radioactive uptake assays, reducing disposal and safety concerns.