Learn More
Molecular Devices Molecular Devices FLIPR Calcium 6-QF Assay Kit, Quench-Free, Sample, 96/384/1536-well
Catalog No. 5000014458
Encompass_Preferred
The Molecular Devices FLIPR Calcium 6-QF Sample Kit is a quench-free, mix-and-read fluorescent reagent set designed for sensitive intracellular calcium flux measurements. The high-quantum-yield dye increases signal amplitude for low-signal targets and the QF formulation enables assay multiplexing while minimizing quenching effects. The sample kit provides reagents sufficient for two microplates and supports 96-, 384-, and 1536-well formats.
  • Quench-free dye formulation enables multiplexing with luminescent assays.
  • High quantum yield increases signal window for low-signal targets.
  • Mix-and-read protocol simplifies workflow and accelerates throughput.
  • Reagents sufficient for two microplates, supporting 96-, 384-, and 1536-well formats.
  • Includes component A (2 vials), component B buffer (1 bottle), and component C (2 vials).
  • Reduces well-to-well variability, improving assay quality and reliability.
Catalog No. 50-000-14458 Supplier Molecular Devices Supplier No. R6134
The Molecular Devices FLIPR Calcium 6-QF Sample Kit is a quench-free, mix-and-read fluorescent reagent set designed for sensitive intracellular calcium flux measurements. The high-quantum-yield dye increases signal amplitude for low-signal targets and the QF formulation enables assay multiplexing while minimizing quenching effects. The sample kit provides reagents sufficient for two microplates and supports 96-, 384-, and 1536-well formats.
  • Quench-free dye formulation enables multiplexing with luminescent assays.
  • High quantum yield increases signal window for low-signal targets.
  • Mix-and-read protocol simplifies workflow and accelerates throughput.
  • Reagents sufficient for two microplates, supporting 96-, 384-, and 1536-well formats.
  • Includes component A (2 vials), component B buffer (1 bottle), and component C (2 vials).
  • Reduces well-to-well variability, improving assay quality and reliability.