Learn More
Molecular Devices Calcium 6-QF Assay Explorer Kit
Supplier:  Molecular Devices R8192/DEL
Encompass_Preferred
Maximizes fluorescent signal from a wide range of cell types by reducing cell wash artifacts. Reduces well-to-well variability, improving assay quality (Z' factor) and reliability (CV %) of high-throughput screens. Universal mix-and-read protocol accelerates assay workflow and increases throughput.
Catalog No. NC0648618