Learn More
Alkali Scientific FlipTop™ 3" Hinged Cardboard Freezer Boxes, 100- Places, White 50/Case
SDP
Catalog No. 503028675
Encompass_Preferred
FlipTop™ hinged lid boxes are made of recyclable cardboard, with a liquid resistant outer coating. The 100-place, 3 inch FlipTop™ box is suitable for tubes up to 12mm in diameter (accepts cryovials up to 4mL). These boxes are shipped fully assembled with dividers. Each box has drain slots on the bottom and numbered grid markings in almost every conceivable location (top, bottom, sides, etc). Also available in 2in. format with various color options. Attached Hinged Lids    Foolproof identification 5 1/4 x 5 1/4 x 3 inches    133 x 133 x 76mm For 81 or 100 tubes    9 x 9 or 10 x 10 format Alpha-numeric coding    Top, sides, and bottom
Catalog No. 50-302-8675 Supplier Alkali Scientific Supplier No. FB3700W
FlipTop™ hinged lid boxes are made of recyclable cardboard, with a liquid resistant outer coating. The 100-place, 3 inch FlipTop™ box is suitable for tubes up to 12mm in diameter (accepts cryovials up to 4mL). These boxes are shipped fully assembled with dividers. Each box has drain slots on the bottom and numbered grid markings in almost every conceivable location (top, bottom, sides, etc). Also available in 2in. format with various color options. Attached Hinged Lids    Foolproof identification 5 1/4 x 5 1/4 x 3 inches    133 x 133 x 76mm For 81 or 100 tubes    9 x 9 or 10 x 10 format Alpha-numeric coding    Top, sides, and bottom