Learn More
Alkali Scientific FlipTop™ Carboard Freezer Box with Attached Hinged Lid, 81-place, Green, 5/cs
SDP
Catalog No. 502389022
Encompass_Preferred
FlipTop Hinged Lid boxes are made of recyclable cardboard, with a liquid resistant outer coating. They are shipped fully assembled with 81-place cardboard dividers. Each box has drain slots on the bottom surface and numbered grid markings in almost every conceivable location (top, bottom, sides, etc). Available in green, pack of five.
Catalog No. 50-238-9022 Supplier Alkali Scientific Supplier No. FB2781G
FlipTop Hinged Lid boxes are made of recyclable cardboard, with a liquid resistant outer coating. They are shipped fully assembled with 81-place cardboard dividers. Each box has drain slots on the bottom surface and numbered grid markings in almost every conceivable location (top, bottom, sides, etc). Available in green, pack of five.