Learn More
MTC Bio Flip Top Vial / Digestion Tube, 50mL, 32x90mm, Self-Standing, Sterile, 20 Bags of 25
Catalog No. NC3737090
Encompass_Preferred
These new flat bottomed vials are made of natural polypropylene. They feature a one-piece design that includes an integral hinged cap and can be used in digestion equipment / protocols.Tubes are supplied in sterile use-packs of 25 pieces, 500 per case.• Self-standing, flat bottom• Attached caps• Molded graduations• Sterile bags of 25 - 500 per case
Catalog No. NC3737090 Supplier MTC Bio Supplier No. T0050
These new flat bottomed vials are made of natural polypropylene. They feature a one-piece design that includes an integral hinged cap and can be used in digestion equipment / protocols.Tubes are supplied in sterile use-packs of 25 pieces, 500 per case.• Self-standing, flat bottom• Attached caps• Molded graduations• Sterile bags of 25 - 500 per case