Learn More
Alkali Scientific FlipTop™ Carboard Freezer Box with Attached Hinged Lid, 100-place, Assorted Colors, 5/cs
SDP
Catalog No. 502335914
Encompass_Preferred
FlipTop Hinged Lid boxes are made of recyclable cardboard, with a liquid resistant outer coating. They are shipped fully assembled with 100-place cardboard dividers. Each box has drain slots on the bottom surface and numbered grid markings in almost every conceivable location (top, bottom, sides, etc). Available in a “Rainbow Pack” of five assorted colors.
Catalog No. 50-233-5914 Supplier Alkali Scientific Supplier No. FB2700A
FlipTop Hinged Lid boxes are made of recyclable cardboard, with a liquid resistant outer coating. They are shipped fully assembled with 100-place cardboard dividers. Each box has drain slots on the bottom surface and numbered grid markings in almost every conceivable location (top, bottom, sides, etc). Available in a “Rainbow Pack” of five assorted colors.