Learn More
Aviva Systems Biology Corp FLJ10490 Antibody - middle region (ARP57114_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57114_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ10490. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FLJ10490. Alternate names: FLJ10490ApplicationWB. Species reactivity: Human
Catalog No. 50-286-266