Learn More
Aviva Systems Biology Corp FLJ13798 Antibody - C-terminal region (ARP33596_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33596_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ13798. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FLJ13798. Alternate names: JMJD5, FLJ13798ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55086