Learn More
Aviva Systems Biology Corp FLJ14213 Antibody - N-terminal region (ARP42637_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42637_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ14213. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FLJ14213. Alternate names: MGC16218, PROTOR2, FLJ14213ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53346