Learn More
Aviva Systems Biology Corp FLJ14768 Antibody - C-terminal region (ARP30009_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30009_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ14768. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human FLJ14768. Alternate names: ZNF798, FLJ14768ApplicationIHC, WB. Species reactivity: Dog, Horse, Human, Pig
Catalog No. 50-014-54096