Learn More
Aviva Systems Biology Corp FLJ20489 Antibody - C-terminal region (ARP42496_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42496_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ20489. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human FLJ20489. Alternate names: HRG1, HRG-1, hHRG-1, FLJ20489ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-58247