Learn More
Aviva Systems Biology Corp FLJ20729 Antibody - C-terminal region (ARP34573_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34573_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ20729. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human FLJ20729. Alternate names: BCD1, C1orf181, NY-BR-75, FLJ20729ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55458