Learn More
Aviva Systems Biology Corp FLJ23436 Antibody - N-terminal region (ARP33583_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33583_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ23436. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FLJ23436. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-55080