Learn More
Aviva Systems Biology Corp FLJ30934 Antibody - middle region (ARP55531_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55531_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ30934. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FLJ30934. Alternate names: DKFZp761P1320, MGC42112, MGC57276, SNX6B, FLJ30934ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-050