Learn More
Aviva Systems Biology Corp FLJ36070 Antibody - N-terminal region (ARP55828_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55828_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ36070. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human FLJ36070. Alternate names: FLJ36070, MASTR, MGC117259ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-087