Learn More
Aviva Systems Biology Corp FLJ36070 Antibody - middle region (ARP55829_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55829_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ36070. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FLJ36070. Alternate names: FLJ36070, MASTR, MGC117259ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-088