Learn More
Aviva Systems Biology Corp FLJ37543 Antibody - middle region (ARP55684_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55684_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ37543. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FLJ37543. Alternate names: MGC138187, MGC138213, FLJ37543ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61578