Learn More
Aviva Systems Biology Corp FLJ43806 Antibody - C-terminal region (ARP62120_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62120_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ43806. It was validated on Western Blot. . Alternate names: KAZ, FLJ10199, FLJ32593, FLJ43398, FLJ43806, KIAA1026, MGC126686, MGC142181, RP1-21O18.1, DKFZp686D08115ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-287-578