Learn More
Foxx Life Sciences EZBiopure Flask Assembly, 500mL, 38-430 mm Cap, PolyCarbonate (PC) Flat Base Flask with, EZBio Tubing and MPC CPC, 8/CS
GREENER_CHOICE
Catalog No. 502558946
Encompass_Preferred
The EZBio Single Use Sterile Erlenmeyer Flask Assembly with TPE Tubing and Vent Filter is designed for fluid transfer and storage. Available in polycarbonate (PC) flask Assemblies or Cap assembly, made with USP Class VI components and Gamma Irradiated for sterility and quality
The EZBio MTO Flask Assemblies allow customized component selection to suit project needs. Customers can choose the bottle volume, male or female connectors, and TPE tubing brand.
Features
Ideal for single use, replacing stainless-steel equipment.
Available in baffled and flat base options with volumes of 1/2L.
Sterility tested to SAL 10-6. Both the flask and VersaCap are autoclavable.
Application
Cell culture incubation
Seed flask inoculation
Bioreactor fed-batch/additions
Media and buffer transfer
Bioprocess Product sampling and supplement injection
Product storage
Filtration and purification process connection
Cell harvesting
Catalog No. 50-255-8946 Supplier Foxx Life Sciences Supplier No. 5689701FLS
The EZBio Single Use Sterile Erlenmeyer Flask Assembly with TPE Tubing and Vent Filter is designed for fluid transfer and storage. Available in polycarbonate (PC) flask Assemblies or Cap assembly, made with USP Class VI components and Gamma Irradiated for sterility and quality
The EZBio MTO Flask Assemblies allow customized component selection to suit project needs. Customers can choose the bottle volume, male or female connectors, and TPE tubing brand.
Features
Ideal for single use, replacing stainless-steel equipment.
Available in baffled and flat base options with volumes of 1/2L.
Sterility tested to SAL 10-6. Both the flask and VersaCap are autoclavable.
Application
Cell culture incubation
Seed flask inoculation
Bioreactor fed-batch/additions
Media and buffer transfer
Bioprocess Product sampling and supplement injection
Product storage
Filtration and purification process connection
Cell harvesting