Learn More
TARGETMOL CHEMICALS INC FLLL32 25MG
SDP
Catalog No. 502576437
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FLLL32 is an effective JAK2/STAT3 inhibitor (IC50 of <5 uM). purity: 99%
Catalog No. 50-257-6437 Supplier TARGETMOL CHEMICALS INC Supplier No. T683825MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FLLL32 is an effective JAK2/STAT3 inhibitor (IC50 of <5 uM). purity: 99%