Learn More
Medchemexpress LLC HY-100544 5mg Medchemexpress, FLLL32 CAS:1226895-15-3 Purity:>98%
SDP
Catalog No. 501931878
Encompass_Preferred
Medchemexpress, HY-100544 5mg FLLL32 CAS:1226895-15-3 FLLL32, a synthetic analog of curcumina, is a JAK2/STAT3 dual inhibitor with anti-tumor activity [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1878 Supplier Medchemexpress LLC Supplier No. HY1005445MG
Medchemexpress, HY-100544 5mg FLLL32 CAS:1226895-15-3 FLLL32, a synthetic analog of curcumina, is a JAK2/STAT3 dual inhibitor with anti-tumor activity [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.