Learn More
O Berk Company FLN SMP JAR 8OZ WHT PP .040 PE
Catalog No. NC9446059
Encompass_Preferred
Sample Jar, 8 oz., Material : Flint Glass, Shape : Round, Neck Thread : GCMI 400, Neck Thread Size : 58 mm
Catalog No. NC9446059 Supplier O Berk Company Supplier No. 28020250
Sample Jar, 8 oz., Material : Flint Glass, Shape : Round, Neck Thread : GCMI 400, Neck Thread Size : 58 mm