Learn More
IDEX Health & Science Upchurch Super Flangeless™ Nut PFA, Natural, 1/4 Flat-Bottom, for 1/16" OD Natural - Single
Catalog No. NC1684509
Encompass_Preferred
This item has a minimum order quantity of 25 per supplier requirements. Super Flangeless™ Nut PFA, Natural, 1/4 Flat-Bottom, for 1/16" OD Natural - Single
Catalog No. NC1684509 Supplier IDEX Health & Science Upchurch Supplier No. P246
This item has a minimum order quantity of 25 per supplier requirements. Super Flangeless™ Nut PFA, Natural, 1/4 Flat-Bottom, for 1/16" OD Natural - Single