Learn More
IDEX Health & Science Upchurch FLNGLSS NUT 1/8IN PTFE 1PK
Catalog No. NC1919834
Encompass_Preferred
Idex Flangeless Nuts: Compatible with 1/4-28, PFA, For Use With: For 0.13 in. O.D. Tubing, Max. Pressure: 500 psi, Flangeless standard knurled head short nut, Certifications/Compliance: ROHS, Natural, Dimensions: 0.87 x 0.37 in. (2.2 x 9.4 cm), Length: 0.87 in., 2.2 cm
Catalog No. NC1919834 Supplier IDEX Health & Science Upchurch Supplier No. P345X
Idex Flangeless Nuts: Compatible with 1/4-28, PFA, For Use With: For 0.13 in. O.D. Tubing, Max. Pressure: 500 psi, Flangeless standard knurled head short nut, Certifications/Compliance: ROHS, Natural, Dimensions: 0.87 x 0.37 in. (2.2 x 9.4 cm), Length: 0.87 in., 2.2 cm