Learn More
Qosina Corp FLO 30™, Tuohy Borst Adapter
Catalog No. NC1291303
Encompass
This item has a minimum qty of 7 per supplier requirements.
Technical Features:0 FR - 9 FR; Up to 11 G (0 inch - 0.118 inch, 0 mm - 3 mm)Materials:PC, Silicone, Teflon
This item is not returnable. View return policy
Catalog No. NC1291303 Supplier Qosina Corp Supplier No. 80330
This item has a minimum qty of 7 per supplier requirements.
Technical Features:0 FR - 9 FR; Up to 11 G (0 inch - 0.118 inch, 0 mm - 3 mm)Materials:PC, Silicone, Teflon