Learn More
Alkali Scientific ASI™ Float Buoys for Floating 3mL Dialysis Chamber, 10/Cs
SDP
Catalog No. 503027275
Encompass_Preferred
ASI™ Float Buoys are available to support a variety of dialysis cassettes for volumes of 0.1 to 20mL.
Catalog No. 50-302-7275 Supplier Alkali Scientific Supplier No. DCSPONGE
ASI™ Float Buoys are available to support a variety of dialysis cassettes for volumes of 0.1 to 20mL.