Learn More
Thomas Scientific Floating Foam Tube Rack, 1.5-2.0mL Tubes, Yellow, 5 pack
SDP
Catalog No. NC1742480
Encompass_Preferred
Brightly-colored, high-density polyethylene foam racks are ideal for floating tubes in water baths, tubs, or beakers. Closed-cell foam resists water absorption so racks can be washed and reused. The blue round rack features unique “X” slits to hold a variety of tube sizes. Racks include carrying handles-easy to lift samples out of baths. Pack of 5.
Catalog No. NC1742480 Supplier Thomas Scientific Supplier No. 1216H49
Brightly-colored, high-density polyethylene foam racks are ideal for floating tubes in water baths, tubs, or beakers. Closed-cell foam resists water absorption so racks can be washed and reused. The blue round rack features unique “X” slits to hold a variety of tube sizes. Racks include carrying handles-easy to lift samples out of baths. Pack of 5.