Learn More
Research Products International Corp Floating Bubble Rack, Round, Capacity 20 7.7mm Tubes
SDP
Catalog No. 50212944
Encompass
Ideal for procedures that require immersion of tubes in a water bath. Bubble racks are molded of foam filled plastic that is injected with small air bubbles for flotation. Sample tubes remain immersed at a constant depth. Racks can be submersed in boiling water and will withstand temperatures up to 175°C.
This item is not returnable. View return policy
Catalog No. 50-212-944 Supplier Research Products International Corp Supplier No. F188751600
Ideal for procedures that require immersion of tubes in a water bath. Bubble racks are molded of foam filled plastic that is injected with small air bubbles for flotation. Sample tubes remain immersed at a constant depth. Racks can be submersed in boiling water and will withstand temperatures up to 175°C.