Learn More
Research Products International Corp Floating Cryotube Rack, Holds 20 1.2 or 2.0ml Cryotubes, 3 3/4 Inch Diameter
SDP
Catalog No. 50212594
Encompass
Floating racks are ideal for quick thawing of frozen cells in cryotubes. Rack floats in waterbaths while samples remain immersed at constant depth regardless of water level. Racks hold 1.2 ml and 2.0 ml cryotubes and can be submerged in boiling water and withstand 135°C. Holes are arranged in a circular pattern on disk. Blue color rack features convenient handle.
Catalog No. 50-212-594 Supplier Research Products International Corp Supplier No. 248699
Add to Cart
Add to Cart
Floating racks are ideal for quick thawing of frozen cells in cryotubes. Rack floats in waterbaths while samples remain immersed at constant depth regardless of water level. Racks hold 1.2 ml and 2.0 ml cryotubes and can be submerged in boiling water and withstand 135°C. Holes are arranged in a circular pattern on disk. Blue color rack features convenient handle.