Learn More
Research Products International Corp Floating Foam Tube Rack, Yellow, Holds 24 1.5 - 2.0ml Tubes, 5 per Package
SDP
Catalog No. 50550321
Encompass
Poly foam floating racks available in three styles for use in water bath applications. Foam racks keep tubes completely submerged. Supplied with optional handles for easy handling.
Catalog No. 50-550-321 Supplier Research Products International Corp Supplier No. 248921
Poly foam floating racks available in three styles for use in water bath applications. Foam racks keep tubes completely submerged. Supplied with optional handles for easy handling.