Learn More
MilliporeSigma™ Floating Rack, Midi
Catalog No. 715133 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 rack
Catalog No. Quantity
71-513-3 10 rack
1 options
Catalog No. 71-513-3 Supplier MilliporeSigma™ Supplier No. 715133
Add to Cart
Add to Cart
Designed to hold D-Tube Dialyzers in an exchange buffer
  • Each Floating Rack, Mini can hold up to nine D-Tube Dialyzer, Mini devices
  • Each Floating Rack, Midi can hold up to four D-Tube Dialyzer, Midi units
  • Each Floating Rack, Maxi can hold up to seven D-Tube Dialyzer, Maxi devices
  • Each Floating Rack, Mega can hold one D-Tube Dialyzer, Mega unit
  • Each package contains 10 Floating Racks
TRUSTED_SUSTAINABILITY
Specifications
Product Type Floating Rack, Midi
Quantity 10 rack
Content And Storage +15°C to +30°C