Learn More
Research Products International Corp Floating Bubble Rack, Round, Capacity 20 10.8mm Tubes
SDP
Catalog No. 50550404
Encompass
Ideal for procedures that require immersion of tubes in a water bath. Bubble racks are molded of foam filled plastic that is injected with small air bubbles for flotation. Sample tubes remain immersed at a constant depth. Racks can be submersed in boiling water and will withstand temperatures up to 175°C.
This item is not returnable. View return policy
Catalog No. 50-550-404 Supplier Research Products International Corp Supplier No. F188751000
Ideal for procedures that require immersion of tubes in a water bath. Bubble racks are molded of foam filled plastic that is injected with small air bubbles for flotation. Sample tubes remain immersed at a constant depth. Racks can be submersed in boiling water and will withstand temperatures up to 175°C.