Learn More
Chemglass Life Sciences Floating Test Tube Rack, Octagon, for PCR, fits 0.25 and 0.5mL Microtubes
SDP
Catalog No. 501897632
Encompass
• Economical and easy to use• Non-absorbentEliminate expensive plastic floating racks with economical polyfoam floating racks. Easy to use in waterbaths or beakers.
Catalog No. 50-189-7632 Supplier Chemglass Life Sciences Supplier No. CGE4770102
• Economical and easy to use• Non-absorbentEliminate expensive plastic floating racks with economical polyfoam floating racks. Easy to use in waterbaths or beakers.