Learn More
Nelson Jameson Company Capitol Vial Floating 24-Vial Rafts
Catalog No. NC1678269
Encompass
All rafts below are manufactured of polycell material. Rafts are flexible and unbreakable allowing a long useful life. Vials fit tightly in holders keeping top and labels out of water. All are 8" x 10".
Catalog No. NC1678269 Supplier Nelson Jameson Company Supplier No. 1703060
All rafts below are manufactured of polycell material. Rafts are flexible and unbreakable allowing a long useful life. Vials fit tightly in holders keeping top and labels out of water. All are 8" x 10".