Learn More
Hach Company Floc Jar
Supplier:  Hach Company 4117000
Encompass
Clear plexiglass, square jar with sample delivery port for monitoring flocculation. Measures 21 x 12.7 x 12.7 cm (8.3 x 5 x 5"), with a capacity of 2 L. Suitable for use with six-unit gang Phipps & Bird or similar floc testing apparatus. Includes tubing, stopper, clamp, and 50 mL graduated beakers.
Catalog No. 50-199-7023