Learn More
Cole-Parmer Stuart Flocculation Jar Tester, 6-place; 230 VAC, 50 Hz
Supplier:  Cole-Parmer 9955105
Encompass_Preferred
These flocculators are designed for repeatable conditions between samples and from run to run. Speed control with electronic feedback accurately maintains the rotational speed of the stainless steel paddles. Two preset programs allow routine speed and time parameters to be entered, stored in memory, and recalled or altered at any time.
Portable tester 99551-00 holds two samples and features a handle for easy transportation. Tester 99551-05 has six positions and features a digital timer. Both testers accept 1-L beakers (not included).
Catalog No. 50-196-1820