Learn More
Formulatrix FLOI8 1000UL TIPS 10 X 96 PCS
SDP
Catalog No. NC2460082
Encompass
Package of FLO i8 F1000 Next Level Tips. Contains 10 x 96 pcs (960 pcs) F1000 conductive pipette tips with filter. Tip working volume from 10 to 1000 µL.
Catalog No. NC2460082 Supplier Formulatrix Supplier No. 800003B
Package of FLO i8 F1000 Next Level Tips. Contains 10 x 96 pcs (960 pcs) F1000 conductive pipette tips with filter. Tip working volume from 10 to 1000 µL.