Learn More
Formulatrix FLOI8 200 UL TIPS 10 X 96 PCS
SDP
Catalog No. NC2460084
Encompass
Package of FLO i8 F200 Next Level Tips. Contains 10 x 96 pcs (960 pcs) F200 conductive pipette tips with filter. Tip working volume from 5 to 200 µL.
Catalog No. NC2460084 Supplier Formulatrix Supplier No. 800002B
Package of FLO i8 F200 Next Level Tips. Contains 10 x 96 pcs (960 pcs) F200 conductive pipette tips with filter. Tip working volume from 5 to 200 µL.