Learn More
Perkin Elmer US LLC Flom PEEK Rotor Seal
Catalog No. 502104586
Encompass_Preferred
PEEK is more inert towards amines and other basic biomolecules/ pharmaceuticals, exhibiting less sample carryover than Vespel. However, Vespel is somewhat more resilient than PEEK, providing better wear life.
Catalog No. 50-210-4586 Supplier Perkin Elmer US LLC Supplier No. N9306068
PEEK is more inert towards amines and other basic biomolecules/ pharmaceuticals, exhibiting less sample carryover than Vespel. However, Vespel is somewhat more resilient than PEEK, providing better wear life.