Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flomoxef sodium | 92823-03-5 | 99.3% | 5 MG
SDP

Catalog No. 5000405320
Encompass_Preferred

Flomoxef sodium is an oxacephem group antibiotic that exhibits excellent activity against various Gram-positive bacteria.

  • Oxacephem group antibiotic
  • Active against Gram-positive bacteria
  • Purity of 99.3%
  • Molecular weight of 518.45
  • Chemical formula: C15H17F2N6NaO7S2
  • White to off-white solid appearance
  • Recommended storage at -20°C, protected from light, under nitrogen.
  • Soluble in DMSO and H2O

Catalog No. 50-004-05320 Supplier Medchemexpress LLC Supplier No. HYB0706A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flomoxef sodium is an oxacephem group antibiotic that exhibits excellent activity against various Gram-positive bacteria.

  • Oxacephem group antibiotic
  • Active against Gram-positive bacteria
  • Purity of 99.3%
  • Molecular weight of 518.45
  • Chemical formula: C15H17F2N6NaO7S2
  • White to off-white solid appearance
  • Recommended storage at -20°C, protected from light, under nitrogen.
  • Soluble in DMSO and H2O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.