Learn More
Medchemexpress LLC Flomoxef sodium | 92823-03-5 | 99.3% | 50 MG
SDP
Catalog No. 5000407967
Encompass_Preferred
Flomoxef sodium is an oxacephem group antibiotic with excellent activity against various Gram-positive bacteria. It has also been investigated for its in-vitro activity against Nocardia. It is a solid, white to off-white compound soluble in DMSO and H2O.
  • Excellent activity against various gram-positive bacteria.
  • In-vitro activity against Nocardia.
  • Solid appearance.
  • White to off-white color.
  • Soluble in DMSO.
  • Soluble in H2O.
Catalog No. 50-004-07967 Supplier Medchemexpress LLC Supplier No. HYB0706A50MG
Flomoxef sodium is an oxacephem group antibiotic with excellent activity against various Gram-positive bacteria. It has also been investigated for its in-vitro activity against Nocardia. It is a solid, white to off-white compound soluble in DMSO and H2O.
  • Excellent activity against various gram-positive bacteria.
  • In-vitro activity against Nocardia.
  • Solid appearance.
  • White to off-white color.
  • Soluble in DMSO.
  • Soluble in H2O.