Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flonicamid-15N,18O | 2483830-60-8 | >98.0% | 120 UG
SDP

Catalog No. 5000233048
Encompass_Preferred

Flonicamid-15N,18O is an isotopically labeled internal standard of the insecticide flonicamid, provided as a methanolic solution for analytical quantification by GC- or LC-MS.

  • Isotopically labeled with 15N and 18O for mass spectrometry accuracy.
  • Supplied as a solution containing 120 μg in 1.2 mL methanol.
  • High purity for analytical use: >98.0%.
  • Molecular weight 232.15 g/mol.
  • Intended as an internal standard for pesticide residue analysis.
  • Safety data sheet and documentation available.

Catalog No. 50-002-33048 Supplier Medchemexpress LLC Supplier No. HY119649S120UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flonicamid-15N,18O is an isotopically labeled internal standard of the insecticide flonicamid, provided as a methanolic solution for analytical quantification by GC- or LC-MS.

  • Isotopically labeled with 15N and 18O for mass spectrometry accuracy.
  • Supplied as a solution containing 120 μg in 1.2 mL methanol.
  • High purity for analytical use: >98.0%.
  • Molecular weight 232.15 g/mol.
  • Intended as an internal standard for pesticide residue analysis.
  • Safety data sheet and documentation available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.