Learn More
Medchemexpress LLC Flonoltinib | 2387765-27-5 | 99.5% | 250 MG
SDP
Catalog No. 5000342624
Encompass_Preferred
Flonoltinib is a potent and orally active dual JAK2/FLT3 inhibitor, exhibiting significant anti-cancer activity. It is intended for research use only.
  • Potent and orally active dual JAK2/FLT3 inhibitor
  • Inhibits JAK2 with an IC50 of 0.7 nM
  • Inhibits FLT3 with an IC50 of 4 nM
  • Inhibits JAK1 with an IC50 of 26 nM
  • Inhibits JAK3 with an IC50 of 39 nM
  • Exhibits significant antitumor effects in vivo
Catalog No. 50-003-42624 Supplier Medchemexpress LLC Supplier No. 250MGHY130247
Flonoltinib is a potent and orally active dual JAK2/FLT3 inhibitor, exhibiting significant anti-cancer activity. It is intended for research use only.
  • Potent and orally active dual JAK2/FLT3 inhibitor
  • Inhibits JAK2 with an IC50 of 0.7 nM
  • Inhibits FLT3 with an IC50 of 4 nM
  • Inhibits JAK1 with an IC50 of 26 nM
  • Inhibits JAK3 with an IC50 of 39 nM
  • Exhibits significant antitumor effects in vivo